Anti-IgG F(c) Rabbit Polyclonal Antibody

Leverancier: Rockland Immunochemicals
ROCK613-4103EA 254 EUR
ROCK613-4103
Anti-IgG F(c) Rabbit Polyclonal Antibody
Antilichamen
Anti-Sheep IgG F(c) antibody has been assayed against 1.0 µg of Sheep IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:200,000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR