Nitrogen generator, Genius 3020
Leverancier: Peak Scientific
PEAK10-6320EA 0 EUR
PEAK10-6320
Nitrogen generator, Genius 3020
Gasgeneratoren Stikstofgeneratoren
The Genius 3020 nitrogen generator combines two individual systems with separate Nitrogen outputs into a single compact unit, thus allowing two separate mass spectrometers to be supplied by the same generator with independent control.
  • Insulated compressor compartment for minimum disruption
  • The gas supplied to both instruments can be switched on and off independently
Maximum Pressure: 6,89 bar
Min/ max operating temperature: 5 to 35 °C
Power consumption: 29900 watts
Weight: 196,5 kg
Noise level: 54 dB
Leveringsinformatie: Supplied with caster wheels for easy mobility.