Anti-NUBPL Mouse Monoclonal Antibody [clone: 5C12]

Leverancier: OriGene
CF503748
ORIGCF503748EA 764 EUR
ORIGCF503748
Anti-NUBPL Mouse Monoclonal Antibody [clone: 5C12]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: NUBPL
Clonality: Monoclonal
Clone: 5C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR