Anti-RBM46 Mouse Monoclonal Antibody [clone: 1E7]
Leverancier: OriGene
CF504551
ORIGCF504551EA 786 EUR
ORIGCF504551
Anti-RBM46 Mouse Monoclonal Antibody [clone: 1E7]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100
Type: Primary
Antigen: RBM46
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human