Anti-C20orf30 Mouse Monoclonal Antibody [clone: 1B4]

Leverancier: OriGene
CF504919
ORIGCF504919EA 764 EUR
ORIGCF504919
Anti-C20orf30 Mouse Monoclonal Antibody [clone: 1B4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: C20orf30
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR