Anti-ATXN7L1 Mouse Monoclonal Antibody [clone: 4F6]

Leverancier: OriGene
CF505708
ORIGCF505708EA 786 EUR
ORIGCF505708
Anti-ATXN7L1 Mouse Monoclonal Antibody [clone: 4F6]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: ATXN7L1
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR