Anti-PDIA4 Mouse Monoclonal Antibody [clone: 2E11]

Leverancier: OriGene
CF503886
ORIGCF503886EA 764 EUR
ORIGCF503886
Anti-PDIA4 Mouse Monoclonal Antibody [clone: 2E11]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PDIA4
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR