Anti-TMEM173 Mouse Monoclonal Antibody [clone: 4H1]

Leverancier: OriGene
CF505023
ORIGCF505023EA 764 EUR
ORIGCF505023
Anti-TMEM173 Mouse Monoclonal Antibody [clone: 4H1]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:100

Type: Primary
Antigen: TMEM173
Clonality: Monoclonal
Clone: 4H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR