Anti-LEPRE1 Mouse Monoclonal Antibody [clone: 1A1]

Leverancier: OriGene
ORIGCF505091EA 764 EUR
ORIGCF505091
Anti-LEPRE1 Mouse Monoclonal Antibody [clone: 1A1]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LEPRE1
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR