Anti-SLA2 Mouse Monoclonal Antibody [clone: 3A7]

Leverancier: OriGene
CF505126
ORIGCF505126EA 764 EUR
ORIGCF505126
Anti-SLA2 Mouse Monoclonal Antibody [clone: 3A7]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: SLA2
Clonality: Monoclonal
Clone: 3A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR