Anti-TRIM45 Mouse Monoclonal Antibody [clone: 2B11]

Leverancier: OriGene
CF505917
ORIGCF505917EA 786 EUR
ORIGCF505917
Anti-TRIM45 Mouse Monoclonal Antibody [clone: 2B11]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: TRIM45
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR