Anti-SENP2 Mouse Monoclonal Antibody [clone: 1C4]

Leverancier: OriGene
CF504810
ORIGCF504810EA 764 EUR
ORIGCF504810
Anti-SENP2 Mouse Monoclonal Antibody [clone: 1C4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SENP2
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR