Anti-TRIM38 Mouse Monoclonal Antibody [clone: 15B10]

Leverancier: OriGene
CF505980
ORIGCF505980EA 764 EUR
ORIGCF505980
Anti-TRIM38 Mouse Monoclonal Antibody [clone: 15B10]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TRIM38
Clonality: Monoclonal
Clone: 15B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR