Anti-ENPEP Mouse Monoclonal Antibody [clone: 3G1]
Leverancier: OriGene
CF504054
ORIGCF504054EA 764 EUR
ORIGCF504054
Anti-ENPEP Mouse Monoclonal Antibody [clone: 3G1]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: ENPEP
Clonality: Monoclonal
Clone: 3G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human