Anti-RAB37 Mouse Monoclonal Antibody [clone: 1E10]

Leverancier: OriGene
CF505339
ORIGCF505339EA 764 EUR
ORIGCF505339
Anti-RAB37 Mouse Monoclonal Antibody [clone: 1E10]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RAB37
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR