Anti-TRIM44 Mouse Monoclonal Antibody [clone: 6F10]

Leverancier: OriGene
CF505510
ORIGCF505510EA 764 EUR
ORIGCF505510
Anti-TRIM44 Mouse Monoclonal Antibody [clone: 6F10]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: TRIM44
Clonality: Monoclonal
Clone: 6F10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR