Anti-TP53I3 Mouse Monoclonal Antibody [clone: 3B11]

Leverancier: OriGene
CF503656
ORIGCF503656EA 786 EUR
ORIGCF503656
Anti-TP53I3 Mouse Monoclonal Antibody [clone: 3B11]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TP53I3
Clonality: Monoclonal
Clone: 3B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR