Anti-PSMD2 Mouse Monoclonal Antibody [clone: 1A12]

Leverancier: OriGene
CF503218
ORIGCF503218EA 764 EUR
ORIGCF503218
Anti-PSMD2 Mouse Monoclonal Antibody [clone: 1A12]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PSMD2
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR