Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 2C3]

Leverancier: OriGene
CF507304
ORIGCF507304EA 786 EUR
ORIGCF507304
Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 2C3]
Antilichamen
Recommended Dilutions: Western Blot: 1:400-4000

Type: Primary
Antigen: ANKMY2
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR