Anti-NEU1 Mouse Monoclonal Antibody [clone: 3C4]

Leverancier: OriGene
CF801668
ORIGCF801668EA 764 EUR
ORIGCF801668
Anti-NEU1 Mouse Monoclonal Antibody [clone: 3C4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NEU1
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR