Anti-PIK3C2A Mouse Monoclonal Antibody [clone: 4E6]

Leverancier: OriGene
CF801710
ORIGCF801710EA 764 EUR
ORIGCF801710
Anti-PIK3C2A Mouse Monoclonal Antibody [clone: 4E6]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PIK3C2A
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR