Anti-RIPK3 Mouse Monoclonal Antibody [clone: 1B3]

Leverancier: OriGene
CF803100
ORIGCF803100EA 786 EUR
ORIGCF803100
Anti-RIPK3 Mouse Monoclonal Antibody [clone: 1B3]
Antilichamen
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: RIPK3
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR