Anti-BIRC3 Mouse Monoclonal Antibody [clone: 3G4]

Leverancier: OriGene
CF800063
ORIGCF800063EA 764 EUR
ORIGCF800063
Anti-BIRC3 Mouse Monoclonal Antibody [clone: 3G4]
Antilichamen
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: BIRC3
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR