Anti-EGFR Mouse Monoclonal Antibody [clone: 1B6]

Leverancier: OriGene
ORIGCF802036EA 764 EUR
ORIGCF802036
Anti-EGFR Mouse Monoclonal Antibody [clone: 1B6]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR