Anti-NUMB Mouse Monoclonal Antibody [clone: 1E3]

Leverancier: OriGene
ORIGCF802140EA 764 EUR
ORIGCF802140
Anti-NUMB Mouse Monoclonal Antibody [clone: 1E3]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NUMB
Clonality: Monoclonal
Clone: 1E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR