Anti-MFN1 Mouse Monoclonal Antibody [clone: 4H4]

Leverancier: OriGene
CF802150
ORIGCF802150EA 764 EUR
ORIGCF802150
Anti-MFN1 Mouse Monoclonal Antibody [clone: 4H4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: MFN1
Clonality: Monoclonal
Clone: 4H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR