Anti-MAP1LC3A Mouse Monoclonal Antibody [clone: 2D5]

Leverancier: OriGene
ORIGCF803407EA 764 EUR
ORIGCF803407
Anti-MAP1LC3A Mouse Monoclonal Antibody [clone: 2D5]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MAP1LC3A
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR