Anti-MSMB Mouse Monoclonal Antibody [clone: 20F2]

Leverancier: OriGene
CF803512
ORIGCF803512EA 764 EUR
ORIGCF803512
Anti-MSMB Mouse Monoclonal Antibody [clone: 20F2]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MSMB
Clonality: Monoclonal
Clone: 20F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR