Anti-PTPRC Mouse Monoclonal Antibody [clone: 2H1]

Leverancier: OriGene
CF506055
ORIGCF506055EA 764 EUR
ORIGCF506055
Anti-PTPRC Mouse Monoclonal Antibody [clone: 2H1]
Antilichamen
Recommended Dilutions: Western Blot: 1:200-8000; Immunochemistry: 1:150; Flow Cytometry: 1:50

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 2H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR