Anti-SIVA1 Mouse Monoclonal Antibody [clone: 2E4]

Leverancier: OriGene
CF506824
ORIGCF506824EA 764 EUR
ORIGCF506824
Anti-SIVA1 Mouse Monoclonal Antibody [clone: 2E4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SIVA1
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR