Anti-WDR74 Mouse Monoclonal Antibody [clone: 4B2]

Leverancier: OriGene
CF800467
ORIGCF800467EA 764 EUR
ORIGCF800467
Anti-WDR74 Mouse Monoclonal Antibody [clone: 4B2]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: WDR74
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR