Anti-PLA2G16 Mouse Monoclonal Antibody [clone: 1B12]

Leverancier: OriGene
CF506914
ORIGCF506914EA 764 EUR
ORIGCF506914
Anti-PLA2G16 Mouse Monoclonal Antibody [clone: 1B12]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PLA2G16
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR