Anti-HMBS Mouse Monoclonal Antibody [clone: 3F4]

Leverancier: OriGene
CF802669
ORIGCF802669EA 764 EUR
ORIGCF802669
Anti-HMBS Mouse Monoclonal Antibody [clone: 3F4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HMBS
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR