Anti-TOLLIP Mouse Monoclonal Antibody [clone: 2E6]

Leverancier: OriGene
ORIGCF507015EA 786 EUR
ORIGCF507015
Anti-TOLLIP Mouse Monoclonal Antibody [clone: 2E6]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: TOLLIP
Clonality: Monoclonal
Clone: 2E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR