Anti-FEN1 Mouse Monoclonal Antibody [clone: 1E1]

Leverancier: OriGene
CF802763
ORIGCF802763EA 764 EUR
ORIGCF802763
Anti-FEN1 Mouse Monoclonal Antibody [clone: 1E1]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FEN1
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR