Anti-FBXW7 Mouse Monoclonal Antibody [clone: 6F5]

Leverancier: OriGene
CF802869
ORIGCF802869EA 764 EUR
ORIGCF802869
Anti-FBXW7 Mouse Monoclonal Antibody [clone: 6F5]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: FBXW7
Clonality: Monoclonal
Clone: 6F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR