Anti-MMP13 Mouse Monoclonal Antibody [clone: 3A7]

Leverancier: OriGene
CF506693
ORIGCF506693EA 764 EUR
ORIGCF506693
Anti-MMP13 Mouse Monoclonal Antibody [clone: 3A7]
Antilichamen
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MMP13
Clonality: Monoclonal
Clone: 3A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR