Anti-TG Mouse Monoclonal Antibody [clone: 8D5]

Leverancier: OriGene
CF804297
ORIGCF804297EA 764 EUR
ORIGCF804297
Anti-TG Mouse Monoclonal Antibody [clone: 8D5]
Antilichamen
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: TG
Clonality: Monoclonal
Clone: 8D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR