Anti-DDX6 Mouse Monoclonal Antibody [clone: 3B7]

Leverancier: OriGene
CF804349
ORIGCF804349EA 764 EUR
ORIGCF804349
Anti-DDX6 Mouse Monoclonal Antibody [clone: 3B7]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DDX6
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR