Anti-PTGS2 Mouse Monoclonal Antibody [clone: 2E1]

Leverancier: OriGene
CF805370
ORIGCF805370EA 764 EUR
ORIGCF805370
Anti-PTGS2 Mouse Monoclonal Antibody [clone: 2E1]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PTGS2
Clonality: Monoclonal
Clone: 2E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR