Anti-NUDT12 Mouse Monoclonal Antibody [clone: 3E7]

Leverancier: OriGene
CF805548
ORIGCF805548EA 786 EUR
ORIGCF805548
Anti-NUDT12 Mouse Monoclonal Antibody [clone: 3E7]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NUDT12
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR