Anti-TGM7 Rabbit Polyclonal Antibody

Leverancier: OriGene
TA320086
ORIGTA320086EA 610 EUR
ORIGTA320086
Anti-TGM7 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: TGM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR