Anti-SLIT2 Rabbit Polyclonal Antibody

Leverancier: OriGene
TA319177
ORIGTA319177EA 860 EUR
ORIGTA319177
Anti-SLIT2 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:1500-1:6000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: SLIT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR