Anti-RNF34 Rabbit Polyclonal Antibody

Leverancier: OriGene
TA319209
ORIGTA319209EA 983 EUR
ORIGTA319209
Anti-RNF34 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:4000-1:20000; Western Blot: 1:500-1:3000

Type: Primary
Antigen: RNF34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR