Anti-HIF1A Rabbit Polyclonal Antibody

Leverancier: OriGene
TA319212
ORIGTA319212EA 860 EUR
ORIGTA319212
Anti-HIF1A Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Monkey, Mouse, Rat, Xenopus
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR