Anti-GDF15 Mouse Monoclonal Antibody [clone: 23B3D2.H5]

Leverancier: OriGene
ORIGTA319547EA 877 EUR
ORIGTA319547
Anti-GDF15 Mouse Monoclonal Antibody [clone: 23B3D2.H5]
Antilichamen
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1:1000

Type: Primary
Antigen: GDF15
Clonality: Monoclonal
Clone: 23B3D2.H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR