Anti-RAD23A Goat Polyclonal Antibody

Leverancier: OriGene
TA319249
ORIGTA319249EA 860 EUR
ORIGTA319249
Anti-RAD23A Goat Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:4000-1:16000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: RAD23A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR