Anti-CCL20 Rat Monoclonal Antibody [clone: 4N5F7]

Leverancier: OriGene
TA319571
ORIGTA319571EA 905 EUR
ORIGTA319571
Anti-CCL20 Rat Monoclonal Antibody [clone: 4N5F7]
Antilichamen
Recommended Dilutions: Western Blot: 0.5?g/ml

Type: Primary
Antigen: CCL20
Clonality: Monoclonal
Clone: 4N5F7
Conjugation: Unconjugated
Epitope: full length
Host: Rat
Isotype:
Reactivity: Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR