Anti-Lefty2 Mouse Monoclonal Antibody [clone: 7C5G1H6H10]

Leverancier: OriGene
TA319606
ORIGTA319606EA 877 EUR
ORIGTA319606
Anti-Lefty2 Mouse Monoclonal Antibody [clone: 7C5G1H6H10]
Antilichamen
Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:2000

Type: Primary
Antigen: Lefty2
Clonality: Monoclonal
Clone: 7C5G1H6H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR