Anti-CD209 Rat Polyclonal Antibody [clone: eB-h209]

Leverancier: OriGene
TA320248
ORIGTA320248EA 377 EUR
ORIGTA320248
Anti-CD209 Rat Polyclonal Antibody [clone: eB-h209]
Antilichamen
Type: Primary
Antigen: CD209
Clonality: Polyclonal
Clone: eB-h209
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR